
Ethyl-2-[(tert-butoxycarbonyl)amino]-1,3-thiazole-5-carboxylate 98%
Catalog # APOSOR5620-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:272.33 g/mol
Melting Pt:198…200 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:302964-01-8
Formula:C₁₁H₁₆N₂O₄S
MDL Number:MFCD06409216