

1-Boc-4-chlorosulphonylpiperidine 98%
Catalog # APOSOR55455-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:283.767 g/mol
Melting Pt:82…85 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
:GHS07
CAS Number:782501-25-1
Formula:C₁₀H₁₈ClNO₄S
MDL Number:MFCD01861750