

5-Aminopentan-1-ol, technická kvalita
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2508-29-4
:
Formula:C₅H₁₃NO
MDL Number:MFCD00008237
Storage Temperature:Ambient
UN:3259
ADR:8,III
:GHS05
:GHS07
Frequently Bought Together