Order Entry
Czech Republic
ContactUsLinkComponent
Diethyl diethylmalonate 98%
Diethyl diethylmalonate 98%
Catalog # APOSOR55121-100G
Supplier:  Apollo Scientific
CAS Number:  
Diethyl diethylmalonate 98%
Catalog # APOSOR55121-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Boiling Pt:228…238 °C
Density:0.99 g/cm³ (25 °C)
Flash Pt:94 °C
 :216.28 g/mol
Storage Temperature:Ambient
CAS Number:77-25-8
Formula:C₁₁H₂₀O₄
MDL Number:MFCD00009132