
1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:98303-20-9
:
Formula:C₁₁H₁₉NO₄
MDL Number:MFCD01862877
:229.276 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together