
3-Chlorobenzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2888-06-4
:
Formula:C₆H₄Cl₂O₂S
MDL Number:MFCD00051697
:211.07 g/mol
Boiling Pt:102…104 °C/1mmHg
Melting Pt:21 °C
Density:1.499 g/cm³ (25 °C)
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:220-753-0
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together