Order Entry
Czech Republic
ContactUsLinkComponent
2-Bromobenzenesulphonyl chloride 95%
Corrosive
2-Bromobenzenesulphonyl chloride 95%
Catalog # APOSOR5370-100G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
2-Bromobenzenesulphonyl chloride 95%
Catalog # APOSOR5370-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :255.52 g/mol
Boiling Pt:117 °C/2.25mmHg
Melting Pt:48…52 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
 :GHS05
CAS Number:2905-25-1
Formula:C₆H₄BrClO₂S
MDL Number:MFCD00051971