
2,7-Dibromophenanthrene-9,10-dione
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:84405-44-7
:
Formula:C₁₄H₆Br₂O₂
MDL Number:MFCD03931078
:366.01 g/mol
Melting Pt:331 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together