Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Bis(3-indenyl)ethane
1,2-Bis(3-indenyl)ethane
1,2-Bis(3-indenyl)ethane
APOSOR52937-5G
:
Apollo Scientific
:
1,2-Bis(3-indenyl)ethane
APOSOR52937-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
258.36 g/mol
:
Ambient
:
18657-57-3
:
C₂₀H₁₈
:
MFCD00191814