Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
D-(+)-Glucosamine hydrochloride 98+%

D-(+)-Glucosamine hydrochloride 98+%

Irritant
Supplier:   Apollo Scientific
D-(+)-Glucosamine hydrochloride 98+%
Supplier:  Apollo Scientific
Certificates
D-(+)-Glucosamine hydrochloride 98+%

3 Options Available Below

Product Details & Documents
Synonyms:  2-Amino-2-deoxy-D-glucopyranose Hydrochloride, D-Glucosamine hydrochloride, (2R,3R,4S,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride
  • CAS Number:66-84-2
  • Formula:C₆H₁₃NO₅
  • MDL Number:MFCD00135831
  • Molecular Weight:215.63 g/mol
  • Melting Pt:∼ 192 °C (dec.)
  • Melting Pt:ca 192 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-638-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 615,00 Kč
    615,00 Kč
    1 * 250 g
       615,00 Kč

  • Bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 1 480,00 Kč
    1 480,00 Kč
    1 * 1 kg
       1 480,00 Kč

  • Bottle
    5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 5 910,00 Kč
    5 910,00 Kč
    1 * 5 kg
       5 910,00 Kč

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.