
2-Di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:564483-19-8
:
Formula:C₂₉H₄₅P
MDL Number:MFCD06411306
:424.65 g/mol
Melting Pt:148…151 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together