Order Entry
Czech Republic
ContactUsLinkComponent
1,4-Dichloropyrido[3,4-d]pyridazine 97%
Irritant
1,4-Dichloropyrido[3,4-d]pyridazine 97%
Catalog # APOSOR52405-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,4-Dichloropyrido[3,4-d]pyridazine 97%
Catalog # APOSOR52405-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :200.0300 g/mol
Melting Pt:159 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:14490-19-8
Formula:C₇H₃Cl₂N₃
MDL Number:MFCD09839782