
(1S,2S)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine 97%
Catalog # APOSOR51933-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:214.31 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:180683-64-1
Formula:C₁₁H₂₂N₂O₂
MDL Number:MFCD08460986