6,7-Dihydro-5H-cyclopenta[b]pyridine
Catalog # APOSOR5187-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:119.17 g/mol
Boiling Pt:87…88 °C/11mmHg
Storage Temperature:Ambient
CAS Number:533-37-9
Formula:C₈H₉N
MDL Number:MFCD00005933