

N-Ethyl-N-2-hydroxyethyl-m-toluidine 96%
Catalog # APOSOR51803-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:179.26 g/mol
Boiling Pt:114…115 °C/0.75mmHg
Density:1.019 g/cm³ (25 °C)
Flash Pt:113 °C
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:91-88-3
Formula:C₁₁H₁₇NO
MDL Number:MFCD00002849