
1,4-Bis(3-aminopropyl)piperazine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7209-38-3
:
Formula:C₁₀H₂₄N₄
MDL Number:MFCD00006161
:200.33 g/mol
Boiling Pt:150…152 °C/2mmHg
Melting Pt:15 °C
Density:0.973 g/cm³ (25 °C)
Flash Pt:163 °C
Storage Temperature:Ambient
UN:2735
ADR:8,II
:GHS05
Frequently Bought Together