
1-[4-(4-Aminophenyl)piperazin-1-yl]ethanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:92394-00-8
:
Formula:C₁₂H₁₇N₃O
MDL Number:MFCD01365904
:219.29 g/mol
Melting Pt:144…148 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together