Order Entry
Czech Republic
ContactUsLinkComponent
4-Bromo-m-toluidine hydrochloride ≥98%
4-Bromo-m-toluidine hydrochloride ≥98%
Catalog # APOSOR510202-100MG
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-m-toluidine hydrochloride ≥98%
Catalog # APOSOR510202-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :222.51
Storage Temperature:Ambient
CAS Number:202925-03-9
Formula:C₇H₉BrClN
MDL Number:MFCD00070748