Order Entry
Czech Republic
ContactUsLinkComponent
2-Chloro-3-methoxybenzonitrile ≥98%
2-Chloro-3-methoxybenzonitrile ≥98%
Catalog # APOSOR510010-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-3-methoxybenzonitrile ≥98%
Catalog # APOSOR510010-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :167.59 g/mol
Storage Temperature:Refrigerator
CAS Number:853331-52-9
Formula:C₈H₆ClNO