
2-Chloronicotinic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2942-59-8
:
Formula:C₆H₄ClNO₂
MDL Number:MFCD00006236
:157.56 g/mol
Melting Pt:176…178 °C (dec.)
EINECS:220-937-0
:GHS07
Storage Temperature:Ambient
Frequently Bought Together