
2,6-Dichlorobenzoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:50-30-6
:
Formula:C₇H₄Cl₂O₂
MDL Number:MFCD00002418
:191.01 g/mol
Melting Pt:139…145 °C
EINECS:200-025-9
:GHS07
Storage Temperature:Ambient
Frequently Bought Together