
1H-Pyrazole 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:288-13-1
:
Formula:C₃H₄N₂
MDL Number:MFCD00005234
Boiling Pt:186…188 °C
Melting Pt:67…70 °C
:68.08 g/mol
Storage Temperature:Ambient
EINECS:206-017-1
:GHS07
Frequently Bought Together