Order Entry
Czech Republic
ContactUsLinkComponent
2-Chloro-5-pyrimidinecarbonyl chloride 95%
Corrosive
2-Chloro-5-pyrimidinecarbonyl chloride 95%
Catalog # APOSOR480739-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
2-Chloro-5-pyrimidinecarbonyl chloride 95%
Catalog # APOSOR480739-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :176.99 g/mol
Storage Temperature:Ambient
UN:1760
ADR:8,III
 :GHS05
CAS Number:110099-99-5
Formula:C₅H₂Cl₂N₂O
MDL Number:MFCD09863165