
3-bromo-4-methoxybenzenesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:23094-96-4
:
Formula:C₇H₆BrClO₃S
MDL Number:MFCD03989643
:285.536 g/mol
Storage Temperature:Ambient
UN:1759
ADR:8,III
:GHS05
Frequently Bought Together