Order Entry
Czech Republic
ContactUsLinkComponent
6-Methoxy-2-naphthylglyoxal hydrate 97%
6-Methoxy-2-naphthylglyoxal hydrate 97%
Catalog # APOSOR480635-5G
Supplier:  Apollo Scientific
CAS Number:  
6-Methoxy-2-naphthylglyoxal hydrate 97%
Catalog # APOSOR480635-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :232.24 g/mol
Storage Temperature:Ambient
CAS Number:745783-88-4
Formula:C₁₃H₁₂O₄
MDL Number:MFCD05664091