
N-(1-Naphthyl)ethylenediamine dihydrochloride 92%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1465-25-4
:
Formula:C₁₂H₁₆Cl₂N₂
MDL Number:MFCD00012556
:259.18 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together