L-Glutamic acid diethyl ester hydrochloride 98%
Catalog # APOSOR480599-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:239.696 g/mol
Storage Temperature:Ambient
CAS Number:1118-89-4
Formula:C₉H₁₈ClNO₄
MDL Number:MFCD00012509