Order Entry
Czech Republic
ContactUsLinkComponent
2,4-Dimethylbenzenesulphonyl chloride 97%
2,4-Dimethylbenzenesulphonyl chloride 97%
Catalog # APOSOR480597-100G
Supplier:  Apollo Scientific
CAS Number:  
2,4-Dimethylbenzenesulphonyl chloride 97%
Catalog # APOSOR480597-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :204.68 g/mol
Storage Temperature:Ambient
CAS Number:609-60-9
Formula:C₈H₉ClO₂S
MDL Number:MFCD00156143