Order Entry
Czech Republic
ContactUsLinkComponent
(3-Methoxyphenyl)hydrazine hydrochloride 98%
(3-Methoxyphenyl)hydrazine hydrochloride 98%
Catalog # APOSOR480591-100G
Supplier:  Apollo Scientific
CAS Number:  
(3-Methoxyphenyl)hydrazine hydrochloride 98%
Catalog # APOSOR480591-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :174.63 g/mol
Storage Temperature:Ambient
CAS Number:39232-91-2
Formula:C₇H₁₁ClN₂O
MDL Number:MFCD00044416