
3,3-Dimethylpiperidine hydrochloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:27832-58-2
:
Formula:C₇H₁₆NCl
MDL Number:MFCD00465824
:149.67 g/mol
Storage Temperature:Ambient
UN:1759
ADR:8,III
:GHS05
Frequently Bought Together