Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(±)-1,2,3,4-Tetrahydro-2-naphthol
(±)-1,2,3,4-Tetrahydro-2-naphthol 98%
(±)-1,2,3,4-Tetrahydro-2-naphthol 98%
APOSOR471677-1G
:
Apollo Scientific
:
(±)-1,2,3,4-Tetrahydro-2-naphthol 98%
APOSOR471677-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
148.2 g/mol
:
GHS07
:
Ambient
:
530-91-6
:
C₁₀H₁₂O
:
MFCD00045575