
Boc-D-4-Cyanophenylalanine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:146727-62-0
:
Formula:C₁₅H₁₈N₂O₄
MDL Number:MFCD00672527
:290.319 g/mol
Melting Pt:152…162 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together