Methyl-5-nitro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6311-73-5
:
Formula:C₆H₅N₃O₆
MDL Number:MFCD01346736
:215.12 g/mol
Storage Temperature:Ambient
Frequently Bought Together