Order Entry
Czech Republic
ContactUsLinkComponent
4,4'-(1E,1'E)-2,2'-(1-Phenyl-1H-pyrazole-3,5-diyl)bis(ethene-2,1-diyl)diphenol
4,4'-(1E,1'E)-2,2'-(1-Phenyl-1H-pyrazole-3,5-diyl)bis(ethene-2,1-diyl)diphenol
Catalog # APOSOR471179-1G
Supplier:  Apollo Scientific
CAS Number:  
4,4'-(1E,1'E)-2,2'-(1-Phenyl-1H-pyrazole-3,5-diyl)bis(ethene-2,1-diyl)diphenol
Catalog # APOSOR471179-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :380.45 g/mol
Storage Temperature:Ambient
CAS Number:828911-79-1
Formula:C₂₅H₂₀N₂O₂
MDL Number:MFCD18433540