Order Entry
Czech Republic
ContactUsLinkComponent
3-Benzylpiperidine-1,3-dicarboxylic acid-1-tert-butyl ester-3-ethyl ester
3-Benzylpiperidine-1,3-dicarboxylic acid-1-tert-butyl ester-3-ethyl ester
Catalog # APOSOR471119-5G
Supplier:  Apollo Scientific
CAS Number:  
3-Benzylpiperidine-1,3-dicarboxylic acid-1-tert-butyl ester-3-ethyl ester
Catalog # APOSOR471119-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :347.46 g/mol
Storage Temperature:Ambient
CAS Number:170842-80-5
Formula:C₂₀H₂₉NO₄
MDL Number:MFCD11500859