Ethyl-2,6-dinitrobenzoate
Catalog # APOSOR471106-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:773136-03-1
Formula:C₉H₈N₂O₆
MDL Number:MFCD06204638