2-Cyanobenzenecarboximidamide hydrochloride
Catalog # APOSOR470985-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:181.63 g/mol
Storage Temperature:Ambient
CAS Number:426828-36-6
Formula:C₈H₈ClN₃
MDL Number:MFCD16622254