Order Entry
Czech Republic
ContactUsLinkComponent
(S)-2-Amino-4-(7-hydroxy-2-oxo-2H-chromen-4-yl)butanoic acid
(S)-2-Amino-4-(7-hydroxy-2-oxo-2H-chromen-4-yl)butanoic acid
Catalog # APOSOR470836-1G
Supplier:  Apollo Scientific
CAS Number:  
(S)-2-Amino-4-(7-hydroxy-2-oxo-2H-chromen-4-yl)butanoic acid
Catalog # APOSOR470836-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :263.249 g/mol
Storage Temperature:Ambient
CAS Number:905442-42-4
Formula:C₁₃H₁₃NO₅
MDL Number:MFCD22377747