Methyl-5-bromo-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate
Catalog # APOSOR470732-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:249.02 g/mol
Storage Temperature:Ambient
CAS Number:30825-88-8
Formula:C₆H₅BrN₂O₄
MDL Number:MFCD07023034