[6-(4-Methoxyphenyl)pyridin-3-yl]methanol
Catalog # APOSOR470280-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:215.25 g/mol
Storage Temperature:Ambient
CAS Number:887974-96-1
Formula:C₁₃H₁₃NO₂
MDL Number:MFCD06410445