Order Entry
Czech Republic
ContactUsLinkComponent
2-Bromo-5-sulphamoylbenzoic acid
2-Bromo-5-sulphamoylbenzoic acid
Catalog # APOSOR470176-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-5-sulphamoylbenzoic acid
Catalog # APOSOR470176-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:22361-61-1
Formula:C₇H₆BrNO₄S
MDL Number:MFCD06355953