Order Entry
Czech Republic
ContactUsLinkComponent
Diethyl (hydroxyimino)malonate
Diethyl (hydroxyimino)malonate
Catalog # APOSOR470144-5G
Supplier:  Apollo Scientific
CAS Number:  
Diethyl (hydroxyimino)malonate
Catalog # APOSOR470144-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :189.17 g/mol
Storage Temperature:Ambient
CAS Number:6829-41-0
Formula:C₇H₁₁NO₅
MDL Number:MFCD00019949