O-1-tert-Butyl-O-3-methyl-(3R)-pyrrolidine-1,3-dicarboxylate
Catalog # APOSOR46748-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:229.28 g/mol
Storage Temperature:Ambient
CAS Number:441717-40-4
Formula:C₁₁H₁₉NO₄
MDL Number:MFCD09954284