
[1,2-Bis(diphenylphosphino)ethane]palladium(II) dichloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:19978-61-1
:
Formula:C₂₆H₂₄Cl₂P₂Pd
MDL Number:MFCD00015702
:575.75 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together