Order Entry
Czech Republic
ContactUsLinkComponent
3-Picolyl bromide hydrobromide 96%
Corrosive
3-Picolyl bromide hydrobromide 96%
Catalog # APOSOR46547-25G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
3-Picolyl bromide hydrobromide 96%
Catalog # APOSOR46547-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :252.94 g/mol
Melting Pt:150…155 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
 :GHS05
CAS Number:4916-55-6
Formula:C₆H₇Br₂N
MDL Number:MFCD01321314