Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 4-methoxyacetoacetate
Methyl-4-methoxyacetoacetate 98%
Methyl-4-methoxyacetoacetate 98%
Methyl-4-methoxyacetoacetate 98%
APOSOR46545-100G
:
Apollo Scientific
:
Methyl-4-methoxyacetoacetate 98%
APOSOR46545-100G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 g
Seq:
3401
:
:
146.1412 g/mol
:
89 °C/8.5mmHg
:
1.129 g/cm³ (25 °C)
:
87 °C
:
GHS07
:
Ambient
:
41051-15-4
:
C₆H₁₀O₄
:
MFCD00010183