


(3S)-(-)-3-(Dimethylamino)pyrrolidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:132883-44-4
:
Formula:C₆H₁₄N₂
MDL Number:MFCD00191348
:114.19 g/mol
Boiling Pt:160…164 °C
Density:0.899 g/cm³ (25 °C)
Flash Pt:51 °C
UN:2734
ADR:8+3,II
:GHS02
:GHS05
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together