Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
N,O-Di-boc-hydroxylamine

N,O-Di-boc-hydroxylamine

Irritant
Supplier:   Apollo Scientific
N,O-Di-boc-hydroxylamine
Supplier:  Apollo Scientific
Certificates
N,O-Di-boc-hydroxylamine

6 Options Available Below

Product Details & Documents
Synonyms:  N,O-Di-Boc-hydroxylamine, tert-Butyl N-(tert-Butoxycarbonyloxy)carbamate, N,O-Bis(tert-butoxycarbonyl)hydroxylamine, tert-Butyl (tert-butoxycarbonyl)oxycarbamate, N-(tert-Butoxycarbonyloxy)carbamic acid tert-Butyl ester
  • CAS Number:85006-25-3
  • Formula:C₁₀H₁₉NO₅
  • MDL Number:MFCD00034797
  • Molecular Weight:233.264 g/mol
  • Melting Pt:67…70 °C
  • Storage Temperature:Freezer
  • EINECS:285-055-0
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 615,00 Kč
    615,00 Kč
    1 * 1 g
       615,00 Kč

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 697,00 Kč
    697,00 Kč
    1 * 5 g
       697,00 Kč

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 1 230,00 Kč
    1 230,00 Kč
    1 * 25 g
       1 230,00 Kč

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 3 490,00 Kč
    3 490,00 Kč
    1 * 100 g
       3 490,00 Kč

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 13 980,00 Kč
    13 980,00 Kč
    1 * 500 g
       13 980,00 Kč

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 68 980,00 Kč
    68 980,00 Kč
    1 * 2,5 kg
       68 980,00 Kč

    Specifications:
    New Product
    New Product