Ethyl-2-hydroxyisobutyrate 98%
Catalog # APOSOR46219-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:132.16 g/mol
Boiling Pt:149…151 °C
Density:0.965 g/cm³ (25 °C)
Flash Pt:41 °C
Storage Temperature:Ambient
CAS Number:80-55-7
Formula:C₆H₁₂O₃
MDL Number:MFCD00004458