
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:250285-32-6
:
Formula:C₂₇H₃₇ClN₂
MDL Number:MFCD02684545
:425.05 g/mol
Melting Pt:278 °C (dec.)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together